Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LECT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204810.1 | NP_990141.1 | 5 - 6 | 616 | 87 | |
GenBank mRNA | AF027380.1 | - | 5 - 6 | 615 | 87 | |
AF138280.1 | - | 5 - 6 | 616 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |