Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDARADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012405.2 | NP_001012405.2 | 3 - 4 | 158 | 141 | |
GenBank mRNA | AJ720393.1 | - | 3 - 4 | 158 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |