Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012543.1 | NP_001012561.1 | 1 - 2 | 158 | 135 | |
GenBank mRNA | AJ851676.1 | - | 1 - 2 | 158 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |