Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012559.1 | NP_001012577.1 | 12 - 13 | 1345 | 95 | |
XM_015281547.2 | XP_015137033.1 | 14 - 15 | 2176 | 95 | ||
XM_015281548.2 | XP_015137034.1 | 13 - 14 | 2173 | 95 | ||
XM_015281549.2 | XP_015137035.1 | 13 - 14 | 1305 | 95 | ||
GenBank mRNA | AJ851567.1 | - | 12 - 13 | 1345 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |