Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCCPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012875.1 | NP_001012893.1 | 11 - 12 | 1256 | 102 | |
GenBank mRNA | AJ719381.1 | - | 11 - 12 | 1256 | 102 | |
CR353828.1 | - | 11 - 12 | 1221 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |