Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012906.1 | NP_001012924.1 | 2 - 3 | 482 | 100 | |
GenBank mRNA | AJ720593.1 | - | 2 - 3 | 482 | 100 | |
CR391018.1 | - | 2 - 3 | 299 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |