Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012917.1 | NP_001012935.1 | 8 - 9 | 1619 | 129 | |
XM_015289441.2 | XP_015144927.2 | 8 - 9 | 1800 | 129 | ||
XM_015289442.2 | XP_015144928.2 | 7 - 8 | 1650 | 129 | ||
XM_015289443.2 | XP_015144929.2 | 7 - 8 | 1536 | 129 | ||
XM_015289444.2 | XP_015144930.2 | 7 - 8 | 1664 | 129 | ||
XM_015289445.2 | XP_015144931.2 | 7 - 8 | 1587 | 129 | ||
XM_025152191.1 | XP_025007959.1 | 8 - 9 | 2079 | 129 | ||
GenBank mRNA | AJ851593.1 | - | 8 - 9 | 1619 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |