Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSFL1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030859.1 | NP_001026030.1 | 7 - 8 | 787 | 128 | |
XM_015296393.2 | XP_015151879.1 | 7 - 8 | 1017 | 128 | ||
GenBank mRNA | AJ720274.1 | - | 7 - 8 | 787 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |