Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001879.1 | NP_001001879.1 | 5 - 6 | 738 | 79 | |
GenBank mRNA | BX932720.1 | - | 5 - 6 | 743 | 79 | |
CR353824.1 | - | 5 - 6 | 738 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |