Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002242.1 | NP_001002242.1 | 7 - 8 | 640 | 93 | |
XM_015296419.2 | XP_015151905.1 | 7 - 8 | 695 | 93 | ||
XM_025142266.1 | XP_024998034.1 | 8 - 9 | 1077 | 93 | ||
XM_025142267.1 | XP_024998035.1 | 7 - 8 | 633 | 93 | ||
XM_025142268.1 | XP_024998036.1 | 7 - 8 | 632 | 93 | ||
XM_025142269.1 | XP_024998037.1 | 7 - 8 | 632 | 93 | ||
XM_025142270.1 | XP_024998038.1 | 7 - 8 | 633 | 93 | ||
GenBank mRNA | M59456.1 | - | 7 - 8 | 640 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |