Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204552.2 | NP_989883.2 | 3 - 4 | 311 | 89 | |
XM_015284410.2 | XP_015139896.1 | 3 - 4 | 492 | 89 | ||
XM_025153522.1 | XP_025009290.1 | 5 - 6 | 789 | 89 | ||
XM_025153546.1 | XP_025009314.1 | 3 - 4 | 444 | 89 | ||
GenBank mRNA | AJ719847.1 | - | 3 - 4 | 311 | 89 | |
AY112660.1 | - | 3 - 4 | 341 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |