Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205016.1 | NP_990347.1 | 2 - 3 | 129 | 75 | |
XM_025141545.1 | XP_024997313.1 | 2 - 3 | 218 | 75 | ||
GenBank mRNA | U79756.1 | - | 2 - 3 | 129 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |