Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205061.1 | NP_990392.1 | 7 | 1009 | 113 | |
GenBank mRNA | Y11030.1 | - | 1009 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Amplicon spans exons and probe does not span exons |