Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205098.1 | NP_990429.1 | 3 - 4 | 177 | 94 | |
XM_015272603.2 | XP_015128089.1 | 2 - 3 | 209 | 94 | ||
XM_015272604.2 | XP_015128090.1 | 2 - 3 | 95 | 94 | ||
XM_015272605.2 | XP_015128091.1 | 2 - 3 | 201 | 94 | ||
GenBank mRNA | AF011356.1 | - | 3 - 4 | 177 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |