Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031174.1 | NP_001026345.1 | 3 - 4 | 296 | 79 | |
XM_015286199.2 | XP_015141685.1 | 3 - 4 | 376 | 79 | ||
XM_025150649.1 | XP_025006417.1 | 3 - 4 | 349 | 79 | ||
GenBank mRNA | AJ719857.1 | - | 3 - 4 | 296 | 79 | |
CR386013.1 | - | 1 - 2 | 55 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |