Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031573.1 | NP_001026744.1 | 4 - 5 | 617 | 134 | |
XM_015282726.2 | XP_015138212.1 | 4 - 5 | 693 | 134 | ||
XM_025147334.1 | XP_025003102.1 | 5 - 6 | 643 | 134 | ||
XM_025147335.1 | XP_025003103.1 | 5 - 6 | 724 | 134 | ||
XM_025147336.1 | XP_025003104.1 | 4 - 5 | 2511 | 134 | ||
XM_025147337.1 | XP_025003105.1 | 6 - 7 | 1369 | 134 | ||
GenBank mRNA | AJ719334.1 | - | 4 - 5 | 617 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |