Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030683.1 | NP_001025854.1 | 3 - 4 | 259 | 101 | |
GenBank mRNA | AJ719730.1 | - | 3 - 4 | 259 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |