Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030715.1 | NP_001025886.1 | 7 - 8 | 1964 | 73 | |
XM_015295747.2 | XP_015151233.1 | 8 - 9 | 2155 | 73 | ||
XM_015295748.2 | XP_015151234.1 | 7 - 8 | 2066 | 73 | ||
XM_015295749.1 | XP_015151235.1 | 6 - 7 | 1860 | 73 | ||
GenBank mRNA | AJ851550.1 | - | 7 - 8 | 1964 | 73 | |
CR391540.1 | - | 5 - 6 | 864 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |