Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030907.1 | NP_001026078.1 | 2 - 3 | 416 | 141 | |
XM_025142999.1 | XP_024998767.1 | 12 - 13 | 6025 | 141 | ||
XM_025143000.1 | XP_024998768.1 | 5 - 6 | 4871 | 141 | ||
XM_025143001.1 | XP_024998769.1 | 12 - 13 | 6014 | 141 | ||
XM_025143002.1 | XP_024998770.1 | 12 - 13 | 6025 | 141 | ||
GenBank mRNA | AJ720328.1 | - | 2 - 3 | 416 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |