Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031356.1 | NP_001026527.1 | 6 - 7 | 1540 | 102 | |
GenBank mRNA | AJ851722.1 | - | 6 - 7 | 1540 | 102 | |
KY982274.1 | - | 6 - 7 | 1305 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |