Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033850.1 | NP_001029022.1 | 2 - 3 | 259 | 147 | |
GenBank mRNA | AB234615.1 | - | 2 - 3 | 259 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |