Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277868.2 | NP_001264797.2 | 7 - 8 | 1263 | 82 | |
NM_001277869.2 | NP_001264798.1 | 7 - 8 | 990 | 82 | ||
XM_025155467.1 | XP_025011235.1 | 7 - 8 | 976 | 82 | ||
XM_025155468.1 | XP_025011236.1 | 7 - 8 | 1027 | 82 | ||
XM_025155469.1 | XP_025011237.1 | 7 - 8 | 891 | 82 | ||
GenBank mRNA | AJ720924.1 | - | 1589 | 82 | ||
CO768688.1 | - | 3 - 4 | 382 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |