Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044635.1 | NP_001038100.1 | 3 - 4 | 380 | 109 | |
XM_015293048.2 | XP_015148534.1 | 4 - 5 | 570 | 109 | ||
XM_015293049.2 | XP_015148535.1 | 4 - 5 | 636 | 109 | ||
XM_015293050.2 | XP_015148536.1 | 4 - 5 | 652 | 109 | ||
XM_015293051.2 | XP_015148537.1 | 4 - 5 | 570 | 109 | ||
GenBank mRNA | AB231851.1 | - | 3 - 4 | 380 | 109 | |
BX932979.1 | - | 4 - 5 | 539 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |