Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL7R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080106.1 | NP_001073575.1 | 2 - 3 | 233 | 72 | |
GenBank mRNA | EF116487.1 | - | 2 - 3 | 233 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |