Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080869.1 | NP_001074338.1 | 2 - 3 | 278 | 145 | |
XM_015282511.2 | XP_015137997.1 | 2 - 3 | 266 | 145 | ||
XM_025147349.1 | XP_025003117.1 | 2 - 3 | 530 | 145 | ||
GenBank mRNA | AJ851481.1 | - | 2 - 3 | 278 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |