Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGDIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080871.1 | NP_001074340.1 | 4 - 5 | 449 | 88 | |
XM_015295424.2 | XP_015150910.1 | 4 - 5 | 443 | 88 | ||
GenBank mRNA | AJ719303.1 | - | 4 - 5 | 449 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |