Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109692.1 | NP_001103162.1 | 4 - 5 | 420 | 102 | |
GenBank mRNA | CR386184.1 | - | 4 - 5 | 391 | 102 | |
CR387537.1 | - | 4 - 5 | 350 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |