Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037275.2 | NP_001032352.2 | 2 - 3 | 1491 | 91 | |
XM_025147326.1 | XP_025003094.1 | 3 - 4 | 1700 | 91 | ||
XM_025147327.1 | XP_025003095.1 | 2 - 3 | 1186 | 91 | ||
XM_025147328.1 | XP_025003096.1 | 2 - 3 | 1186 | 91 | ||
GenBank mRNA | AM087669.1 | - | 2 - 3 | 215 | 91 | |
DQ117917.1 | - | 2 - 3 | 586 | 91 | ||
DQ278876.1 | - | 2 - 3 | 1491 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |