Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204100.1 | NP_989431.1 | 6 - 7 | 605 | 95 | |
XM_015287950.2 | XP_015143436.1 | 5 - 6 | 587 | 95 | ||
GenBank mRNA | AB021166.1 | - | 5 - 6 | 590 | 95 | |
AB059657.1 | - | 6 - 7 | 605 | 95 | ||
AJ719978.1 | - | 6 - 7 | 619 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |