Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204199.1 | NP_989530.1 | 6 - 7 | 631 | 116 | |
XM_015280581.2 | XP_015136067.1 | 6 - 7 | 653 | 116 | ||
XM_015280582.2 | XP_015136068.1 | 6 - 7 | 653 | 116 | ||
GenBank mRNA | AF322650.1 | - | 6 - 7 | 631 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |