Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204203.2 | NP_989534.1 | 4 - 5 | 334 | 78 | |
XM_015293984.1 | XP_015149470.1 | 3 - 4 | 285 | 78 | ||
XM_025154780.1 | XP_025010548.1 | 3 - 4 | 620 | 78 | ||
GenBank mRNA | AF190051.1 | - | 4 - 5 | 336 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |