Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204213.1 | NP_989544.1 | 4 - 5 | 723 | 78 | |
XM_015292118.2 | XP_015147604.1 | 5 - 6 | 857 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |