Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204217.1 | NP_989548.1 | 3 - 4 | 303 | 61 | |
GenBank mRNA | AY215074.1 | - | 3 - 4 | 303 | 61 | |
BX930857.2 | - | 3 - 4 | 290 | 61 | ||
BX933819.1 | - | 525 | 61 | |||
X07257.1 | - | 248 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |