Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204279.1 | NP_989610.1 | 6 - 7 | 1823 | 84 | |
XM_015276625.2 | XP_015132111.1 | 6 - 7 | 1865 | 84 | ||
GenBank mRNA | AF075160.1 | - | 6 - 7 | 1823 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |