Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204334.1 | NP_989665.1 | 3 - 4 | 676 | 116 | |
GenBank mRNA | AF329195.1 | - | 3 - 4 | 676 | 116 | |
CR385995.1 | - | 4 - 5 | 714 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |