Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204440.1 | NP_989771.1 | 1 - 2 | 425 | 85 | |
GenBank mRNA | AY163159.1 | - | 1 - 2 | 425 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |