Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205205.2 | NP_990536.2 | 3 - 4 | 647 | 79 | |
XM_015281306.2 | XP_015136792.1 | 4 - 5 | 650 | 79 | ||
XM_015281307.2 | XP_015136793.1 | 4 - 5 | 650 | 79 | ||
GenBank mRNA | CR382426.1 | - | 3 - 4 | 647 | 79 | |
X80258.1 | - | 577 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |