Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205284.1 | NP_990615.1 | 7 - 8 | 898 | 90 | |
XM_015286580.2 | XP_015142066.1 | 7 - 8 | 909 | 90 | ||
GenBank mRNA | X53828.1 | - | 898 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |