Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006194.1 | NP_001006194.1 | 6 - 7 | 846 | 141 | |
XM_015275331.2 | XP_015130817.1 | 6 - 7 | 1065 | 141 | ||
XM_015275332.2 | XP_015130818.1 | 6 - 7 | 1035 | 141 | ||
XM_015275333.2 | XP_015130819.1 | 6 - 7 | 995 | 141 | ||
GenBank mRNA | AJ719719.1 | - | 6 - 7 | 846 | 141 | |
BX933900.2 | - | 5 - 6 | 808 | 141 | ||
BX935621.1 | - | 6 - 7 | 836 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |