Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006223.1 | NP_001006223.1 | 4 - 5 | 871 | 78 | |
XM_015295773.2 | XP_015151259.2 | 6 - 7 | 5839 | 78 | ||
XM_015295774.2 | XP_015151260.2 | 5 - 6 | 867 | 78 | ||
GenBank mRNA | AJ719792.1 | - | 4 - 5 | 871 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |