Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006274.1 | NP_001006274.1 | 5 - 6 | 1256 | 152 | |
GenBank mRNA | AJ720484.1 | - | 5 - 6 | 1256 | 152 | |
AY332740.1 | - | 5 - 6 | 1256 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |