Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006300.1 | NP_001006300.1 | 5 - 6 | 400 | 95 | |
XM_015296855.2 | XP_015152341.1 | 4 - 5 | 364 | 95 | ||
GenBank mRNA | AJ721109.1 | - | 5 - 6 | 400 | 95 | |
CR387498.1 | - | 4 - 5 | 380 | 95 | ||
CR390617.1 | - | 5 - 6 | 401 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |