Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD79B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006328.2 | NP_001006328.2 | 1 - 2 | 168 | 76 | |
GenBank mRNA | AJ719814.1 | - | 1 - 2 | 168 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |