Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006489.1 | NP_001006489.1 | 3 - 4 | 429 | 121 | |
XM_015287481.2 | XP_015142967.1 | 3 - 4 | 433 | 121 | ||
XM_025150795.1 | XP_025006563.1 | 2 - 3 | 279 | 121 | ||
GenBank mRNA | AJ719558.1 | - | 3 - 4 | 429 | 121 | |
BX935798.2 | - | 3 - 4 | 430 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |