Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006491.1 | NP_001006491.1 | 5 - 6 | 471 | 59 | |
GenBank mRNA | AJ719752.1 | - | 5 - 6 | 471 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |