Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007905.2 | NP_001007906.1 | 4 - 5 | 705 | 89 | |
GenBank mRNA | BX933632.1 | - | 4 - 5 | 441 | 89 | |
BX933635.1 | - | 4 - 5 | 458 | 89 | ||
BX933691.1 | - | 4 - 5 | 439 | 89 | ||
BX935045.2 | - | 4 - 5 | 468 | 89 | ||
BX935881.1 | - | 4 - 5 | 445 | 89 | ||
BX935913.1 | - | 4 - 5 | 701 | 89 | ||
BX950755.2 | - | 4 - 5 | 439 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |