Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DESI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008460.1 | NP_001008460.1 | 2 - 3 | 474 | 122 | |
XM_025148721.1 | XP_025004489.1 | 13 - 14 | 4496 | 122 | ||
XM_025148722.1 | XP_025004490.1 | 12 - 13 | 4413 | 122 | ||
GenBank mRNA | AJ720677.1 | - | 2 - 3 | 474 | 122 | |
BX930740.1 | - | 2 - 3 | 127 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |