Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004400.2 | NP_001004400.1 | 4 | 1548 | 118 | |
GenBank mRNA | V00389.1 | - | 1548 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |