Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130585.1 | NP_001124057.1 | 1 - 2 | 360 | 83 | |
GenBank mRNA | EU732608.1 | - | 1 - 2 | 360 | 83 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Gallus gallus Probe spans exons |