Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010842.2 | NP_001010842.2 | 1 | 945 | 71 | |
GenBank mRNA | AB154518.1 | - | 1 | 855 | 71 | |
BX933448.2 | - | 1 | 945 | 71 | ||
BX933520.2 | - | 1 | 945 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Inventoried Both primers and probe map within a single exon |